missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11420750
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
10031-20-6
Br2H8MnO4
286.81
MFCD00149791
HHDPJRSXPOGIOP-UHFFFAOYSA-L
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
manganese(2+) tetrahydrate dibromide
O.O.O.O.[Mn++].[Br-].[Br-]
This item is not returnable. View return policy
10031-20-6
Br2H8MnO4
286.81
MFCD00149791
HHDPJRSXPOGIOP-UHFFFAOYSA-L
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
manganese(2+) tetrahydrate dibromide
O.O.O.O.[Mn++].[Br-].[Br-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 64.3°C (decomposition)
Color Pink
Physical Form Crystalline
Quantity 50 g
Merck Index 14,5725
Formula Weight 214.76 (Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Manganese (II) bromide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'