missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) carbonate hydrate, 90% Product Code.: 10636222

Manganese(II) carbonate hydrate, 90%

Product Code. 10636222
1 kg, Plastic bottle
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10636222

Brand: Thermo Scientific Acros 222540010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34156-69-9 | CMnO3 | 114.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 34156-69-9
Physical Form Amorphous Powder
Quantity 1 kg
Assay Percent Range 88% min. (Complexometry)
Molecular Formula CMnO3
Linear Formula MnCO3·xH2O
MDL Number MFCD00149794
Synonym manganese ii carbonate hydrate, co3.mn.h2o, manganese 2+ hydrate carbonate, manganese 2+ ion hydrate carbonate
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute inorganic acid, insoluble in alcohol, practically insoluble in common organic acids
InChI Key XMWCXZJXESXBBY-UHFFFAOYSA-L
SMILES [Mn++].[O-]C([O-])=O
IUPAC Name manganese(2+) carbonate
Molecular Weight (g/mol) 114.95
PubChem CID 24848673
Formula Weight 114.95
Percent Purity 90%
Loss on Drying 3% max. (105°C)
Packaging Plastic bottle
Specific Gravity 3.12
Density 3.1200g/mL
Chemical Name or Material Manganese(II) carbonate hydrate, 90%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461874 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog Number222540010
Lot NumberA0400953 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number222540010
Lot NumberA0403267 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number222540010
Lot NumberA0417144 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number222540010
Lot NumberA0442142 Certificate TypeCertificate of Analysis Date16/05/2022 Catalog Number222540010
Product Identifier
  • Manganese(II) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) carbonate hydrate, 90% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.