Learn More
Manganese(II) chloride, 1M aq. soln., Thermo Scientific Chemicals
£47.77
Chemical Identifiers
CAS | 7773-01-5 |
---|---|
Molecular Formula | Cl2Mn |
Molecular Weight (g/mol) | 125.84 |
MDL Number | MFCD00011114 |
InChI Key | GLFNIEUTAYBVOC-UHFFFAOYSA-L |
Synonym | manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystallineShow More |
PubChem CID | 10313134 |
ChEBI | CHEBI:63041 |
IUPAC Name | manganese(2+) dichloride |
SMILES | [Cl-].[Cl-].[Mn++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15405419
|
Thermo Scientific Alfa Aesar
J63150.AD |
50 mL |
£47.77
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15415419
|
Thermo Scientific Alfa Aesar
J63150.AE |
100 mL |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7773-01-5 | |
125.84 | |
GLFNIEUTAYBVOC-UHFFFAOYSA-L | |
10313134 | |
manganese(2+) dichloride |
Cl2Mn | |
MFCD00011114 | |
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystallineShow More | |
CHEBI:63041 | |
[Cl-].[Cl-].[Mn++] |
Specifications
7773-01-5 | |
Liquid | |
MnCl2 | |
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline | |
[Cl-].[Cl-].[Mn++] | |
125.84 | |
CHEBI:63041 | |
1M aq. soln. |
Colorless | |
Cl2Mn | |
MFCD00011114 | |
GLFNIEUTAYBVOC-UHFFFAOYSA-L | |
manganese(2+) dichloride | |
10313134 | |
125.84 | |
Manganese(II) chloride |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P260-P280-P305+P351+P338-P310-P314-P501c
H318-H373
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Manganese chloride, 1M aqueous solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Manganese(II) chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.