missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) chloride tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10552991

Manganese(II) chloride tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10552991
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10552991

Brand: Thermo Scientific Acros 205895000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58.0°C
Color Pink
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula MnCl2·4H2O
Fieser 07,222; 09,288
Merck Index 15,5793
Solubility Information Solubility in water: 1980g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 197.91
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Manganese(II) chloride tetrahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0471712 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog Number205890025,205895000,205891000
Lot NumberA0464270 Certificate TypeCertificate of Analysis Date11/07/2024 Catalog Number205890025,205895000,205891000
Lot NumberA0459787 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog Number205890025,205895000,205891000
Lot NumberA0453359 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number205890025,205895000,205891000
Lot NumberA0395906 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number205890025,205895000,205891000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) chloride tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) chloride tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.