Learn More
Manganese(II) chloride tetrahydrate, 99% (metals basis), Thermo Scientific Chemicals
£30.25 - £108.54
Chemical Identifiers
CAS | 13446-34-9 |
---|---|
Molecular Formula | Cl2H8MnO4 |
Molecular Weight (g/mol) | 197.90 |
MDL Number | MFCD00149792 |
InChI Key | CNFDGXZLMLFIJV-UHFFFAOYSA-L |
IUPAC Name | manganese(2+) tetrahydrate dichloride |
SMILES | O.O.O.O.[Cl-].[Cl-].[Mn++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347287
|
Thermo Scientific Alfa Aesar
011563.22 |
100 g |
£30.25
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357287
|
Thermo Scientific Alfa Aesar
011563.36 |
500 g |
£108.54
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Manganese(II) chloride tetrahydrate is used in the tailing of DNA - MnCl2 buffer is recommended for dC and dG tailing and in reverse transcription of RNA with thermostable DNA polymerase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13446-34-9 | |
197.90 | |
CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
O.O.O.O.[Cl-].[Cl-].[Mn++] |
Cl2H8MnO4 | |
MFCD00149792 | |
manganese(2+) tetrahydrate dichloride |
Specifications
58°C | |
Pink | |
99% (Metal basis) | |
MFCD00149792 | |
CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
manganese(2+) tetrahydrate dichloride | |
197.90 (125.84 Anhydrous) | |
Hygroscopic | |
2.010 g/cm3 at 20°C |
13446-34-9 | |
Crystalline | |
Cl2H8MnO4 | |
Soluble in pyridine,ethanol. Insoluble in ether. | |
O.O.O.O.[Cl-].[Cl-].[Mn++] | |
197.90 | |
Odorless | |
Pink Crystalline/powder | |
Manganese(II) chloride tetrahydrate |
Safety and Handling
P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
H302-H318-H373
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 231-869-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Manganese(II) chloride tetrahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Manganous chloride tetrahydrate
RUO – Research Use Only