Learn More
Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Plays an important role in the production of dry cell batteries.
£31.46 - £416.07
Chemical Identifiers
CAS | 7773-01-5 |
---|---|
Molecular Formula | Cl2Mn |
Molecular Weight (g/mol) | 125.84 |
MDL Number | MFCD00011114 |
InChI Key | GLFNIEUTAYBVOC-UHFFFAOYSA-L |
Synonym | manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline |
PubChem CID | 10313134 |
ChEBI | CHEBI:63041 |
IUPAC Name | manganese(2+);dichloride |
SMILES | [Cl-].[Cl-].[Mn++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11330068
|
Thermo Scientific Alfa Aesar
042844.04 |
2 g |
£31.46
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320068
|
Thermo Scientific Alfa Aesar
042844.09 |
10 g |
£113.93
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11310068
|
Thermo Scientific Alfa Aesar
042844.18 |
50 g |
£416.07
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7773-01-5 | |
125.84 | |
GLFNIEUTAYBVOC-UHFFFAOYSA-L | |
10313134 | |
manganese(2+);dichloride |
Cl2Mn | |
MFCD00011114 | |
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline | |
CHEBI:63041 | |
[Cl-].[Cl-].[Mn++] |
Specifications
1190°C | |
7773-01-5 | |
Cl2Mn | |
MFCD00011114 | |
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline | |
GLFNIEUTAYBVOC-UHFFFAOYSA-L | |
manganese(2+);dichloride | |
10313134 | |
125.84 | |
Ampouled under argon | |
2.977 | |
Manganese(II) chloride |
650°C | |
Powder | |
MnCl2 | |
UN3288 | |
Soluble in water,pyridine and alcohol. Insoluble in ether. | |
[Cl-].[Cl-].[Mn++] | |
125.84 | |
CHEBI:63041 | |
99.99% | |
Hygroscopic | |
2.977 |
Safety and Handling
P262-P264-P270-P280i-P301+P312-P305+P351+P338-P310-P314-P330-P501c
H302-H318-H373
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 231-869-6
missing translation for 'rtecsNumber' : OO9625000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.