Learn More
Manganese(II) fluoride, 99%, Thermo Scientific Chemicals
£38.09 - £120.60
Chemical Identifiers
CAS | 7782-64-1 |
---|---|
Molecular Formula | F2Mn |
Molecular Weight (g/mol) | 92.935 |
MDL Number | MFCD00016222 |
InChI Key | CTNMMTCXUUFYAP-UHFFFAOYSA-L |
Synonym | manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis |
PubChem CID | 24528 |
IUPAC Name | difluoromanganese |
SMILES | F[Mn]F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397207
|
Thermo Scientific Alfa Aesar
011527.09 |
10 g |
£38.09
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387207
|
Thermo Scientific Alfa Aesar
011527.18 |
50 g |
£120.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Manganese(II) fluoride is used in the manufacture of special kinds of glass and lasers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-64-1 | |
92.935 | |
CTNMMTCXUUFYAP-UHFFFAOYSA-L | |
24528 | |
F[Mn]F |
F2Mn | |
MFCD00016222 | |
manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis | |
difluoromanganese |
Specifications
856°C | |
Red | |
99% | |
MFCD00016222 | |
Insoluble in water,alcohol. Soluble in dilute HF,conc. HCl or HNO3 | |
F[Mn]F | |
92.935 | |
92.93 | |
3.98 |
7782-64-1 | |
Crystalline Powder | |
F2Mn | |
manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis | |
CTNMMTCXUUFYAP-UHFFFAOYSA-L | |
difluoromanganese | |
24528 | |
Hygroscopic | |
Manganese(II) fluoride |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-960-0
missing translation for 'rtecsNumber' : OP0875000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only