missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) fluoride, Puratronic, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15407395
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Manganese(II) fluoride is used in the production of special kinds of glass and lasers. It reacts with antimony pentafluoride in hydrofluoric acid to prepare a complex, Mn(SbF6)2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-64-1
Molecular Formula F2Mn
Molecular Weight (g/mol) 92.935
MDL Number MFCD00016222
InChI Key CTNMMTCXUUFYAP-UHFFFAOYSA-L
Synonym manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis
PubChem CID 24528
IUPAC Name difluoromanganese
SMILES F[Mn]F

Specifications

Melting Point 856°C
Color Red
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula MnF2
UN Number UN3288
Merck Index 14,5729
Solubility Information Soluble in dilute hydrogen fluoride,concentrated hydrochloric acid and nitric acid; Insoluble in water and alcohol.
Formula Weight 92.93
Percent Purity 99.99%
Grade Puratronic™
Sensitivity Hygroscopic
Density 3.98 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(II) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'