missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) iodide, anhydrous, 98%

Product Code. 11373018
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11373018

Brand: Thermo Scientific Alfa Aesar 032808.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-33-2

Manganese(II) iodide can be used as a pink pigment or as a source of the manganese ion or iodide ion. It is often used in the lighting industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-33-2
MDL Number MFCD00016225

Specifications

Melting Point 638°C
Color Red to White
Physical Form Powder
Quantity 50 g
Linear Formula MnI2
Merck Index 14,5732
Solubility Information Soluble in water with gradual decomposition; Soluble in alcohol
Formula Weight 308.75
Percent Purity 98%
Odor Odorless
Sensitivity Air, light, and moisture sensitive
Density 5.0 g/mL
Chemical Name or Material Manganese(II) iodide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.