missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11347668
2 kg
Click to view available options
Quantity:
50 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Manganese(II) nitrate is used as a precursor to prepare the oxides of manganese. It is involved in the preparation of electronic components. Further, it is utilized for the phosphorous treatment of metal surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-66-9
Molecular Formula MnN2O6
Molecular Weight (g/mol) 178.946
MDL Number MFCD00011112
InChI Key MIVBAHRSNUNMPP-UHFFFAOYSA-N
Synonym manganese nitrate, manganous nitrate, manganese dinitrate, manganous dinitrate, manganese ii nitrate, manganese 2+ nitrate, nitric acid, manganese 2+ salt, manganese ii nitrate, anhydrous, manganesenitrate
PubChem CID 61511
IUPAC Name manganese(2+);dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2]

Specifications

Color Pink
Physical Form Liquid
Quantity 2 kg
Solubility Information Miscible with water.
Formula Weight 178.95
Appearance Pink liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Manganese(II) nitrate, 50% w/w aq. Soln

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253153 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number033340.A3,033340.36
Lot Number10252190 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog Number033340.A3,033340.36
Lot Number10241795 Certificate TypeCertificate of Analysis Date01/07/2022 Catalog Number033340.A3,033340.36
Lot Number10239917 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number033340.A3,033340.36
Lot NumberK07L34 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number033340.A3,033340.36
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) nitrate, 50% w/w aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Manganese nitrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) nitrate, 50% w/w aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.