missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

£19.40 - £203.01

Chemical Identifiers

CAS 15710-66-4
Molecular Formula MnN2O6
Molecular Weight (g/mol) 178.95
MDL Number MFCD00149788
InChI Key MIVBAHRSNUNMPP-UHFFFAOYSA-N
Synonym manganese ii nitrate hydrate, acmc-1brrd, manganese nitrate hydrate, mn.2no3.h2o, manganese 2+ ion hydrate dinitrate, manganese 2+ hydrate dinitrate, manganese ii nitrate hydrate, puratronic, manganese 2+ nitrate-water 1/2/1Show MoreShow Less
PubChem CID 16211480
SMILES [Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11356917
SDS View Documents
Thermo Scientific Alfa Aesar
010806.04
2 g
£19.40
2g
Please to purchase this item. Need a web account? Register with us today!
11366917
SDS View Documents
Thermo Scientific Alfa Aesar
010806.09
10 g
£53.27
10g
Please to purchase this item. Need a web account? Register with us today!
11376917
SDS View Documents
Thermo Scientific Alfa Aesar
010806.18
50 g
£203.01
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Raw material for electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. Also useful precursors to the oxides of manganese.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H272-H302-H314-H373

missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III

missing translation for 'einecsNumber' : 233-828-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.