missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) nitrate tetrahydrate, 98%

Product Code. p-7024206
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11488437

Brand: Thermo Scientific Alfa Aesar A18521.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20694-39-7 | H8MnN2O10 | 251.006 g/mol

Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

Solubility
Soluble in ethanol and water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong reducing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20694-39-7
Molecular Formula H8MnN2O10
Molecular Weight (g/mol) 251.006
MDL Number MFCD00149789
InChI Key ALIMWUQMDCBYFM-UHFFFAOYSA-N
Synonym manganese ii nitrate tetrahydrate, manganese nitrate tetrahydrate, unii-03m0g68r5f, manganese 2+ ion tetrahydrate dinitrate, manganese dinitrate tetrahydrate, acmc-20akkk, nitric acid, manganese 2+ salt, tetrahydrate, ksc206g2f, mn.2no3.4h2o, manganese 2+ dinitrate tetrahydrate
PubChem CID 182616
IUPAC Name manganese(2+);dinitrate;tetrahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Mn+2]

Specifications

Melting Point ∼26°C
Quantity 250 g
Linear Formula Mn(NO3)2·4H2O
UN Number UN2724
Merck Index 14,5733
Solubility Information Soluble in ethanol and water.
Formula Weight 251.01 (178.95 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Manganese(II) nitrate tetrahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) nitrate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Manganese(II) nitrate tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.