missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals

Product Code. 11306998
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

As a paint and varnish drier

Manganese(II) oxalate dihydrate is used as a paint and varnish drier. It is also used as a chemical reagent as well as to prepare other manganese compounds. In addition, it is used as a semiconductor photosensitive material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-16-7
Molecular Formula C2H4MnO6
Molecular Weight (g/mol) 178.986
MDL Number MFCD00150452
InChI Key HDJUVFZHZGPHCQ-UHFFFAOYSA-L
Synonym manganese ii oxalate 2-hydrate
PubChem CID 131882857
IUPAC Name manganese(2+);oxalate;dihydrate
SMILES C(=O)(C(=O)[O-])[O-].O.O.[Mn+2]

Specifications

Melting Point 100°C (decomposition)
Color White
Density 2.453 g/mL
Odor Odorless
Quantity 5 g
Merck Index 14,5734
Solubility Information Soluble in dilute acids; Slightly soluble in water.
Formula Weight 178.99 (142.96 Anhydrous)
Concentration or Composition (by Analyte or Components) Mn 30% min.
Physical Form Chunks and/or Powder
Chemical Name or Material Manganese(II) oxalate dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'