Learn More
Manganese(II) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Description
The main use of manganese dioxide is in dry-cell batteries, such as the alkaline battery and the zinc-carbon battery. It is useful as a pigment, and as a precursor to other manganese compounds. It is used as a reagent in organic synthesis, for instance, for the oxidation of allylic alcohols and arylmethyl alcohols. It is also employed in various industrial purposes such as adhesives and sealant chemicals, lubricants and lubricant additives, fertilizers, oxidizing/reducing agents, plating agents and surface treating agents, processing aids, air care products, paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Green |
Physical Form | Chunks and/or Powder |
Quantity | 10 g |
Solubility Information | Soluble in hydrochloric acid. Insoluble in water. |
Formula Weight | 70.94 |
Percent Purity | 99.99% |
Odor | Odorless |
Density | 5.0 g/mL |
Assay | (metals basis) |
Chemical Name or Material | Manganese (II) oxide |
RUO – Research Use Only