missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11396917
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11396917

Brand: Thermo Scientific Alfa Aesar 010807.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 15244-36-7 | H2MnO5S | 169.01 g/mol

Used in feed additive, nutrient, firming agent and flavor enhancer. It is also used as a fermentation aid in the processing of beer and malt beverages.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-36-7
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Color Pink
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99.999% (Metal basis)
IUPAC Name manganese(2+);sulfate;hydrate
Formula Weight 151.00 (Anhydrous)
Odor Odorless
Appearance Pink Crystalline Powder
Chemical Name or Material Manganese(II) sulfate hydrate, Loses all water at 400-450°

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.