missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(Ii) Sulfate Monohydrate, ≥98%, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | MnSO4 · H2O |
UN Number | UN3077 |
IUPAC Name | manganese(2+);sulfate;hydrate |
Formula Weight | 169.02g/mol |
Percent Purity | ≥98% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Chemical Name or Material | Manganese(Ii) Sulfate Monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'