missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) sulfate monohydrate, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10696592

Manganese(II) sulfate monohydrate, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10696592
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10696592

Brand: Thermo Scientific Acros 205905000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrateShow MoreShow Less
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 850.0°C
Melting Point 700.0°C
Color Pink to White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula MnSO4·H2O
Merck Index 15, 5804
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 169.02
Percent Purity ≥99%
Grade Extra Pure
Loss on Ignition 10.0% to 12.0% (500°C)
Packaging Plastic bottle
Chemical Name or Material Manganese(II) sulfate monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460382 Certificate TypeCertificate of Analysis Date27/02/2024 Catalog Number205900010,205900050,205905000
Lot NumberA0455088 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number205900010,205900050,205905000
Lot NumberA0399777 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number205900010,205900050,205905000
Lot NumberA0410587 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number205900010,205900050,205905000
Lot NumberA0445958 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog Number205900010,205900050,205905000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) sulfate monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Manganese sulfate monohydrate, Manganese(II) sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) sulfate monohydrate, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.