missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Sulfate Monohydrate, ≥99%, Honeywell™, Reagentplus™
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | 700°C |
Color | Red |
pH | 3.0 to 3.5 (at 50g/L 20°C) |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | MnSO4 · H2O |
UN Number | UN3077 |
Formula Weight | 169.02g/mol |
Percent Purity | ≥99% |
Packaging | Poly bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'