missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) sulfate monohydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur., 99.0-101.0%
£46.21 - £1860.00
Chemical Identifiers
CAS | 10034-96-5 |
---|---|
Molecular Formula | H2MnO5S |
Molecular Weight (g/mol) | 169.01 |
MDL Number | MFCD00149159 |
InChI Key | ISPYRSDWRDQNSW-UHFFFAOYSA-L |
Synonym | manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate |
PubChem CID | 177577 |
ChEBI | CHEBI:86364 |
IUPAC Name | manganese(2+);sulfate;hydrate |
SMILES | O.[Mn++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15613400
|
Honeywell-Fluka
31425-500G |
500 g | N/A | ||||||
15603400
|
Honeywell-Fluka
31425-1KG |
1 kg | N/A | ||||||
15633400
|
Honeywell-Fluka
31425-6X1KG |
6 x 1 kg | N/A | ||||||
15623400
|
Honeywell-Fluka
31425-50KG |
50 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10034-96-5 | |
169.01 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
177577 | |
manganese(2+);sulfate;hydrate |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
CHEBI:86364 | |
O.[Mn++].[O-]S([O-])(=O)=O |
Specifications
10034-96-5 | |
MnSO4 · H2O | |
3077 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
manganese(2+);sulfate;hydrate | |
177577 | |
169.02g/mol | |
ACS/puriss. p.a. |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
O.[Mn++].[O-]S([O-])(=O)=O | |
169.01 | |
CHEBI:86364 | |
99 to 101% | |
Manganese(Ii) Sulfate Monohydrate, Reag. Ph. Eur. |
Safety and Handling
P273
H373-H411
missing translation for 'einecsNumber' : 232-089-9
missing translation for 'rtecsNumber' : OP0893500
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Manganese(II) sulfate monohydrate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Manganese(II) sulfate-1-hydrate