missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Sulfate, Pure, Dried Powder, Fisher Chemical
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Specifications
Boiling Point | 850°C |
Melting Point | 700°C |
CAS | 10034-96-5 |
Color | White |
pH | 3 to 3.5 |
Physical Form | Powder/Solid |
Quantity | 1 kg |
Molecular Formula | H2MnO5S |
MDL Number | MFCD00149159 |
Synonym | manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate |
Show More |
Safety and Handling
Product Identifier
- Manganese(II) sulfate monohydrate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'