missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) sulfate tetrahydrate, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 11489577

Manganese(II) sulfate tetrahydrate, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11489577
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11489577

Brand: Thermo Scientific Alfa Aesar B22081.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.

Solubility
Soluble in water.

Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-68-5
Molecular Formula H8MnO8S
Molecular Weight (g/mol) 223.054
MDL Number MFCD00149160
InChI Key CDUFCUKTJFSWPL-UHFFFAOYSA-L
Synonym manganese sulfate tetrahydrate, manganese ii sulfate tetrahydrate, unii-f46lh60l4m, manganese sulphate, tetrahydrate, mnso4.4h2o, acmc-1c6k2, mn.so4.4h2o, manganese 2+ sulfate tetrahydrate, manganese ii sulphate tetrahydrateShow MoreShow Less
PubChem CID 22572548
ChEBI CHEBI:86358
IUPAC Name manganese(2+);sulfate;tetrahydrate
SMILES O.O.O.O.[O-]S(=O)(=O)[O-].[Mn+2]

Specifications

Quantity 100 g
Linear Formula MnSO4·4H2O
UN Number UN3077
Solubility Information Soluble in water.
Formula Weight 223.06 (151.00 Anhydrous)
Percent Purity 99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Manganese(II) sulfate tetrahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250169 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog NumberB22081.36,B22081.22
Lot Number10250170 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog NumberB22081.36,B22081.22
Lot Number10243418 Certificate TypeCertificate of Analysis Date09/09/2022 Catalog NumberB22081.36,B22081.22
Lot Number10189296 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22081.36,B22081.22
Lot Number10154656 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22081.36,B22081.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) sulfate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid, manganese(2+) salt tetrahydrate, Manganese(II) sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) sulfate tetrahydrate, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.