missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Sulfate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Description
Manganese(II) Sulfate Tetrahydrate, Certified AR for Analysis
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
Melting Point | 26°C to 27°C |
Color | Red |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 223.05g/mol |
Packaging | Plastic powder jar |
Sécurité et traitement
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Identifiant produit
- Manganese(II) sulfate, tetrahydrate
- Danger
- Specific target organ toxicity after repeated exposure Category 2
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Manganese(II) sulfate, Manganese(II) sulfate tetrahydrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'