missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Sulfate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7073291 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
This item is not returnable. View return policy

Product Code. 11452844

Brand: Fisher Chemical M/2300/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manganese(II) Sulfate Tetrahydrate, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-68-5
Molecular Formula MnO4S
Molecular Weight (g/mol) 150.99
MDL Number MFCD00149159
InChI Key SQQMAOCOWKFBNP-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+) sulfate
SMILES [Mn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 26°C to 27°C
Color Red
Physical Form Solid
Quantity 250 g
Formula Weight 223.05g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) sulfate, tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Manganese(II) sulfate, Manganese(II) sulfate tetrahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.