missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

£85.50 - £373.86

Identificadores de productos químicos

CAS 12032-74-5
Molecular Formula MnTiO3
MDL Number MFCD00016216
Synonym manganese titanium trioxide, manganese ii titanate, manganese 2+ ; oxygen 2-; titanium 4+, titanium 4+ ion manganese 2+ ion trioxidandiide
Ver más especificaciones

Productos 2
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
11382228
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
013133.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11372228
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
013133.18
50 g
£373.86
50g
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Manganese(II) titanium oxide is used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Especificaciones

Identificadores de productos químicos

12032-74-5
MnTiO3
MFCD00016216
manganese titanium trioxide, manganese ii titanate, manganese 2+ ; oxygen 2-; titanium 4+, titanium 4+ ion manganese 2+ ion trioxidandiide

Especificaciones

1360°C
Yellow
99.9% (Metals basis)
MFCD00016216
Insoluble in water.
Odorless
Manganese(II) titanium oxide
12032-74-5
Powder
MnTiO3
manganese titanium trioxide, manganese ii titanate, manganese 2+ ; oxygen 2-; titanium 4+, titanium 4+ ion manganese 2+ ion trioxidandiide
150.84
4.54g/cm3 at 20°C
Videos
Seguridad y manipulación

Seguridad y manipulación

missing translation for 'einecsNumber' : 234-776-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documentos

Documentos

Certificados

RUO – Research Use Only