Learn More
Manganese(III) acetate dihydrate, 97%, Thermo Scientific Chemicals
£109.00 - £597.00
Chemical Identifiers
CAS | 19513-05-4 |
---|---|
Molecular Formula | C6H9MnO6 |
Molecular Weight (g/mol) | 232.07 |
MDL Number | MFCD00150022 |
InChI Key | AHSBSUVHXDIAEY-UHFFFAOYSA-K |
IUPAC Name | manganese(3+) triacetate |
SMILES | [Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402837
|
Thermo Scientific Alfa Aesar
047050.04 |
2 g |
£109.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412837
|
Thermo Scientific Alfa Aesar
047050.09 |
10 g |
£165.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15422837
|
Thermo Scientific Alfa Aesar
047050.18 |
50 g | |||||||
Description
Manganese(III) acetate dihydrate, is used as a mild and selective oxidizing agent. It is also a reagent used for radical cyclizations and α-keto-acetoxylation. Mangese triacetate has been used as a single electron oxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19513-05-4 | |
232.07 | |
AHSBSUVHXDIAEY-UHFFFAOYSA-K | |
[Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
C6H9MnO6 | |
MFCD00150022 | |
manganese(3+) triacetate |
Specifications
19513-05-4 | |
Solid | |
Mn(OOCCH3)3·2H2O | |
3732626 | |
AHSBSUVHXDIAEY-UHFFFAOYSA-K | |
manganese(3+) triacetate | |
268.10 (232.07 Anhydrous) | |
Hygroscopic |
Pink | |
C6H9MnO6 | |
MFCD00150022 | |
It is soluble in water. | |
[Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
232.07 | |
97% | |
Manganese(III) acetate dihydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
H315-H319-H335-H336-H373
missing translation for 'einecsNumber' : 213-602-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only