Learn More
Manganese(III) fluoride, 98% (metals basis), Thermo Scientific Chemicals
£54.77 - £191.96
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322168
|
Thermo Scientific Alfa Aesar
013069.09 |
10 g |
£54.77
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332168
|
Thermo Scientific Alfa Aesar
013069.18 |
50 g |
£191.96
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a fluorinating agent, useful for conversion of hydrocarbons into fluorocarbons. It is used in organic synthesis as Lewis acid to prepare a number of organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-53-1 | |
MFCD00016223 |
Specifications
(decomposition) | |
Crystalline | |
UN3288 | |
Insoluble in organic solvents. Hydrolyzed by water. | |
98% | |
3.54 g/mL | |
Manganese(III) fluoride |
7783-53-1 | |
MFCD00016223 | |
14,5741 | |
111.93 | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H272-H302+H332-H315-H318-H335
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 232-006-6
missing translation for 'rtecsNumber' : OP0882600
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only