missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(IV) oxide, 80-85%, powder, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 535.0°C |
Color | Gray |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 80-85% |
Linear Formula | MnO2 |
Fieser | 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202 |
Merck Index | 15, 5795 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid |
Formula Weight | 86.94 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'