missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 97%, Thermo Scientific Chemicals

£24.42 - £545.72

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunsteinShow MoreShow Less
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488830
SDS View Documents
Thermo Scientific Alfa Aesar
A10765.09
10 g
£24.42
10g
Please to purchase this item. Need a web account? Register with us today!
11469662
SDS View Documents
Thermo Scientific Alfa Aesar
A10765.36
500 g
£58.59
500g
Please to purchase this item. Need a web account? Register with us today!
11479662
SDS View Documents
Thermo Scientific Alfa Aesar
A10765.0E
2500 g
£183.92
2500g
Please to purchase this item. Need a web account? Register with us today!
15463027
SDS View Documents
Thermo Scientific Alfa Aesar
A10765.A9
10 kg
£545.72
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.

Solubility
Insoluble in water.

Notes
Incompatible with strong acids, strong reducing agents and organic materials.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(IV) oxide, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.