missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 99%, -10 mesh, Thermo Scientific Chemicals

Product Code. 11974141
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535.0°C
Quantity 2.5 kg
Assay Percent Range 99%
Linear Formula MnO2
UN Number 1479
Fieser 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202
Merck Index 15,5795
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid
Formula Weight 86.94
Percent Purity 99%
Packaging Plastic Bottle
Density 5.02
Chemical Name or Material Manganese(IV) Oxide, 99+%, -100 mesh
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'