missing translation for 'onlineSavingsMsg'
Learn More
Manganese(IV) oxide, 99%, -10 mesh, Thermo Scientific Chemicals Product Code.: 11994141

Manganese(IV) oxide, 99%, -10 mesh, Thermo Scientific Chemicals

Product Code. 11994141
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 11994141

Brand: Thermo Scientific Acros 222585000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535.0°C
Quantity 500 g
Assay Percent Range 99%
Linear Formula MnO2
UN Number 1479
Fieser 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202
Merck Index 15,5795
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid
Formula Weight 86.94
Percent Purity 99%
Packaging Plastic bottle
Density 5.02
Chemical Name or Material Manganese(IV) oxide, 99+%, -10 mesh
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460073 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number222580025,222581000,222585000
Lot NumberA0370827 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number222580025,222581000,222585000
Lot NumberA0417804 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number222580025,222581000,222585000
Lot NumberA0447858 Certificate TypeCertificate of Analysis Date08/12/2022 Catalog Number222580025,222581000,222585000
Lot NumberA0439683 Certificate TypeCertificate of Analysis Date07/03/2022 Catalog Number222580025,222581000,222585000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(IV) oxide, 99%, -10 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.