missing translation for 'onlineSavingsMsg'
Learn More

Manganese phosphide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11351549
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Manganese phosphide is used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12263-33-1
Molecular Formula Mn3P2
Molecular Weight (g/mol) 226.76
MDL Number MFCD00064736
InChI Key ZZDYLDYDQHMVJZ-UHFFFAOYSA-N
IUPAC Name trimanganese(2+) diphosphanetriide
SMILES [P-3].[P-3].[Mn++].[Mn++].[Mn++]

Specifications

Melting Point 1095°C
Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 226.75
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Manganese phosphide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'