missing translation for 'onlineSavingsMsg'
Learn More
Manganese, plasma standard solution, Specpure™ Mn 10,000μg/mL, Thermo Scientific Chemicals Product Code.: 11362348

Manganese, plasma standard solution, Specpure™ Mn 10,000μg/mL, Thermo Scientific Chemicals

Product Code. 11362348
100 mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11362348

Brand: Thermo Scientific Alfa Aesar 014413.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011111

Specifications

Quantity 100 mL
UN Number UN3264
Physical Form Liquid
Concentration Mn in 5% HNO3
Grade Specpure™
Chemical Name or Material Manganese plasma standard solution, Mn 10,000μg/mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1544582 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number014413.AP,014413.AD,014413.AE
Lot Number1542172 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog Number014413.AP,014413.AD,014413.AE
Lot Number1528005 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog Number014413.AP,014413.AD,014413.AE
Lot Number1518884 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number014413.AP,014413.AD,014413.AE
Lot Number1524496 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number014413.AP,014413.AD,014413.AE
compliance-icons
Product Identifier
  • Manganese, plasma standard solution, Specpure, Mn 10,000µg/m
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese, plasma standard solution, Specpure™ Mn 10,000μg/mL, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.