Learn More
Manganese powder, -140+325 mesh, 99.6% (metals basis), Thermo Scientific Chemicals
£29.65 - £272.36
Chemical Identifiers
CAS | 7439-96-5 |
---|---|
Molecular Formula | Mn |
Molecular Weight (g/mol) | 54.94 |
MDL Number | MFCD00011111 |
InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
Synonym | colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds |
PubChem CID | 23930 |
ChEBI | CHEBI:35154 |
IUPAC Name | manganese |
SMILES | [Mn] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334058
|
Thermo Scientific Alfa Aesar
041343.14 |
25 g |
£29.65
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314058
|
Thermo Scientific Alfa Aesar
041343.22 |
100 g |
£77.59
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324058
|
Thermo Scientific Alfa Aesar
041343.36 |
500 g |
£272.36
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-96-5 | |
54.94 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
23930 | |
manganese |
Mn | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compoundsShow More | |
CHEBI:35154 | |
[Mn] |
Specifications
7439-96-5 | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
[Mn] | |
54.94 | |
CHEBI:35154 | |
Powder | |
Manganese powder |
Mn | |
UN3089 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
manganese | |
23930 | |
99.6% | |
Metals basis |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 231-105-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.