missing translation for 'onlineSavingsMsg'
Learn More
Manganese powder, -325 mesh, 99.3% (metals basis), Thermo Scientific Chemicals Product Code.: 11301308

Manganese powder, -325 mesh, 99.3% (metals basis), Thermo Scientific Chemicals

Product Code. 11301308
25 g
Click to view available options
Quantity:
25 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11301308

Brand: Thermo Scientific Alfa Aesar 010238.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Apparent density 2.6-3.5g/cm3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-96-5
Molecular Formula Mn
Molecular Weight (g/mol) 54.94
MDL Number MFCD00011111
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
PubChem CID 23930
ChEBI CHEBI:35154
IUPAC Name manganese
SMILES [Mn]

Specifications

Color Gray
Appearance Gray powder
Assay Percent Range 99.3% (Metals basis)
Quantity 25 g
Mesh Size -325 Mesh
Form Powder
Chemical Name or Material Manganese

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62500037 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number010238.A1,010238.30,010238.14
Lot NumberQ20I040 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number010238.A1,010238.30,010238.14
Lot NumberY18H033 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010238.A1,010238.30,010238.14
Lot NumberA29L21 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010238.A1,010238.30,010238.14
Lot NumberD08P02 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010238.A1,010238.30,010238.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese powder, -325 mesh, 99.3% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.