missing translation for 'onlineSavingsMsg'
Learn More

Manganese solution for ICP 10g/L HCl matrix, Fisher Chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical S 233 500

Code : 04

Additional Details : Weight : 0.50000kg Transport : UN number : 3264 Chem class : 8 Pack group : III


Product Code. 12916344

  • £362.00 / 500mL
Estimated Shipment: 23-09-2024
to see stock.

Description

Description

Specifications

Specifications

Ampoule
EU Ph Ref: 1050501
Malachite green solution
1000 mL
Eu. Ph.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Single-element Standard Solution for ICP. Manganese (Mn) 10000mg/l in HCl 5% (S 233)

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers