missing translation for 'onlineSavingsMsg'
Learn More
Manganese sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™ Product Code.: 15424805

Manganese sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™

Product Code. 15424805
1 Ea.
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy

Product Code. 15424805

Brand: Thermo Scientific Alfa Aesar 040865.KS

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-96-5
Molecular Formula Mn
MDL Number MFCD00011111
Quantity 1 Ea.
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
SMILES [Mn]
IUPAC Name manganese
Molecular Weight (g/mol) 54.94
PubChem CID 23930
ChEBI CHEBI:35154
Percent Purity 99.95%
Odor Odorless
Form Hot Pressed
Assay (metals basis)
Chemical Name or Material Manganese sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.