missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannose triflate, 98%, Thermo Scientific Chemicals
£42.10 - £245.00
Chemical Identifiers
CAS | 92051-23-5 |
---|---|
Molecular Formula | C15H19F3O12S |
Molecular Weight (g/mol) | 480.36 |
InChI Key | OIBDVHSTOUGZTJ-PEBLQZBPSA-N |
Synonym | tatm, mannose triflate, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-beta-d-mannopyranose, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-b-d-mannopyranose, unii-0ki8ffq6tt, 0ki8ffq6tt, beta-d-mannopyranose 1,3,4,6-tetra-o-acetate 2-o-trifluoromethanesulfonate, 2r,3r,4s,5s,6s-3,4,6-tris acetyloxy-5-trifluoromethanesulfonyloxy oxan-2-yl methyl acetate, mannose trilfate |
PubChem CID | 11113597 |
IUPAC Name | [(2R,3R,4S,5S,6S)-3,4,6-triacetyloxy-5-(trifluoromethylsulfonyloxy)oxan-2-yl]methyl acetate |
SMILES | CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OS(=O)(=O)C(F)(F)F)OC(=O)C)OC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15587049
|
Thermo Scientific Acros
461341000 |
100 mg |
£42.10
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15577049
|
Thermo Scientific Acros
461340010 |
1 g |
£245.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
92051-23-5 | |
480.36 | |
tatm, mannose triflate, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-beta-d-mannopyranose, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-b-d-mannopyranose, unii-0ki8ffq6tt, 0ki8ffq6tt, beta-d-mannopyranose 1,3,4,6-tetra-o-acetate 2-o-trifluoromethanesulfonate, 2r,3r,4s,5s,6s-3,4,6-tris acetyloxy-5-trifluoromethanesulfonyloxy oxan-2-yl methyl acetate, mannose trilfate | |
[(2R,3R,4S,5S,6S)-3,4,6-triacetyloxy-5-(trifluoromethylsulfonyloxy)oxan-2-yl]methyl acetate |
C15H19F3O12S | |
OIBDVHSTOUGZTJ-PEBLQZBPSA-N | |
11113597 | |
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OS(=O)(=O)C(F)(F)F)OC(=O)C)OC(=O)C |
Specifications
92051-23-5 | |
Authentic | |
C15H19F3O12S | |
Clear colorless to slightly yellow | |
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OS(=O)(=O)C(F)(F)F)OC(=O)C)OC(=O)C | |
480.36 | |
480.36 | |
Mannose triflate |
118.0°C to 122.0°C | |
97.5% min. (ELSD) (HPLC) | |
tatm, mannose triflate, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-beta-d-mannopyranose, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-b-d-mannopyranose, unii-0ki8ffq6tt, 0ki8ffq6tt, beta-d-mannopyranose 1,3,4,6-tetra-o-acetate 2-o-trifluoromethanesulfonate, 2r,3r,4s,5s,6s-3,4,6-tris acetyloxy-5-trifluoromethanesulfonyloxy oxan-2-yl methyl acetate, mannose trilfate | |
OIBDVHSTOUGZTJ-PEBLQZBPSA-N | |
[(2R,3R,4S,5S,6S)-3,4,6-triacetyloxy-5-(trifluoromethylsulfonyloxy)oxan-2-yl]methyl acetate | |
11113597 | |
98% |
RUO – Research Use Only