Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Brand: Thermo Scientific™ C4032-102
343.73 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Place decrimping head squarely on seal and squeeze.- Minimizes damage to vial rim with chromatography or serum vials
- Removes standard aluminum seals and many steel seals
Specifications
Industrial Chromatography | |
National | |
Manual Decrimper | |
1 Ea. |
Aluminum And Steel Seal, National Vial, Chromacol Vial | |
32 mm | |
Manual Decrimper |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.