Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Use Thermo Scientific™ Manual Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
£306.00 - £394.00
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
17314163
|
Thermo Scientific™
60180-CR11 |
11 mm Crimp Vials | Manual Crimper |
£394.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17324163
|
Thermo Scientific™
60180-CR13 |
13 mm Crimp Vials | Manual Crimper |
£382.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17334163
|
Thermo Scientific™
60180-CR20 |
20 mm Crimp Vials | Manual Crimper |
£382.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17354163
|
Thermo Scientific™
60180-CR8 |
8 mm Crimp Vials | Manual Crimper |
£306.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17364163
|
Thermo Scientific™
60180-DCR11 |
11 mm Crimp Vials | Manual Decrimper |
£342.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17374163
|
Thermo Scientific™
60180-DCR13 |
13 mm Crimp Vials | Manual Decrimper |
£332.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17383763
|
Thermo Scientific™
60180-DCR20 |
20 mm Crimp Vials | Manual Decrimper |
£394.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
Specifications
Stainless Steel/Aluminum | |
SureSTART Level 2 |