missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Manual Decapper
£207.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10064610
|
Macherey-Nagel™
735408 |
8 mm Aluminum Caps |
£207.00
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10144661
|
Macherey-Nagel™
735911 |
11 mm Aluminum Crimp Caps |
£207.00
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
Specifications
Manual Decapper | |
1 Each |