missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Manual Decapper, Steel, Non-autoclavable
Manual decrimping tools allow easy removal of aluminum seals without breakage.
Brand: Fisherbrand™ 13 07 0003
206.08 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
- Decapper
- Manual
- Steel
Specifications
Aluminum Cap | |
Manual | |
13mm Decapper |