missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Manual Flask Handler
Brand: BUCHI 041414
Code : 62
Additional Details : Weight : 3.80000kg
Specifications
Manual Flask Handler | |
For 50L Flask | |
1 Pack | |
50 L |