Learn More
SGE Analytical Science™ Manual Syringe
Improve the sensitivity of your MS analysis with this syringe. SGE Analytical Science™ Manual Syringes feature individually fitted plungers for a perfect seal.
Brand: SGE Analytical Science™ 004050
This item is not returnable.
View return policy
Description
Improve the sensitivity of your MS analysis with this syringe. SGE Analytical Science™ Manual Syringes feature individually fitted plungers for a perfect seal.
- Removable Needle with standard plunger
- Lowest syringe carryover
- Reduced non-specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment
Specifications
25 G | |
50 mm | |
50 μL | |
Bevel |
0.50 mm | |
Manual Syringe | |
SGE | |
Removable |