missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAOB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12602-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The MAOB gene encodes a 520-amino acid protein with a molecular mass of 58.8 kDa and shows 70% amino acid identity to MAOA. MAOA and MAOB are present in the outer mitochondrial membrane in the central nervous system and peripheral tissues. A comparison of highly purified human placental MAOA and human liver MAOB revealed that the A form of the enzyme is larger by 2 kDa and only one potential N-glycosylation site exists in each protein, with each site in a different relevant position.Specifications
MAOB | |
Polyclonal | |
Unconjugated | |
MAOB | |
MAO B, MAOB, Monoamine Oxidase B, Monoamine oxidase type B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109731, 25750, 4129 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P19643, P27338, Q8BW75 | |
MAOB | |
MAOB Fusion Protein Ag3253 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |