missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPK10 (JNK3) Recombinant Human Protein
MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs).
Brand: Invitrogen™ PR6983B
This item is not returnable.
View return policy
Description
MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4.
Specifications
NP_002744 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100 μg | |
RUO - research use only | |
MAPK10 (JNK3) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
76.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MAP Kinase Family | |
Recombinant | |
Baculovirus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction