missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPK12 (p38 γ) Recombinant Human Protein, Inactive
MAPK12 (p38 gamma) is a serine/threonine kinase involved in many different cellular processes.
Brand: Invitrogen™ PV3653
Code : 50
Additional Details : Weight : 0.05250kg
Description
MAPK12 (p38 gamma) is activated during hypoxia and gamma-irradiation-induced G(2) arrest.
Specifications
NP_002960 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
46kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length, Recombinant, Inactive | |
mitogen-activated protein kinase 12 | |
Human | |
His Tag | |
Kinases & Inhibitors |
Cell Analysis, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation, Cell Signaling, Kinase Assay, Enzyme and Protein Activity Assay, Industrial and Applied Science, Pharma and Biopharma, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Cell Proliferation and Function | |
6300 | |
100μg | |
RUO - research use only | |
MAPK12 (p38 γ) | |
MAP Kinase Family | |
Recombinant | |
Baculovirus | |
Serine/Threonine Kinases |