Learn More
MAPK14 & EGFR Protein Protein Interaction Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DI0027.1Set
This item is not returnable.
View return policy
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAPK14 protein, and the other against the EGFR protein for use in in situ Proximity Ligation Assay.Specifications
MAPK14 | |
Unconjugated | |
CSBP1/CSBP2/CSPB1/EXIP/Mxi2/PRKM14/PRKM15/RK/SAPK2A/p38/p38ALPHA | |
1 Set | |
Primary | |
Antibody pair set content: 1. MAPK14 rabbit purified polyclonal antibody (20 ug) 2. EGFR mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
In situ PLA | |
mitogen-activated protein kinase 14 | |
MAPK14 | |
RUO | |
1432/1956 | |
Human | |
Ab Pair |
Product Content Correction
Product Title
MAPK14 & EGFR Protein Protein Interaction Antibody Pair, Abnova™ > 1 Set; Unconjugated
Spot an opportunity for improvement?Share a Content Correction