Learn More
MARCH5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12213-1-AP-150UL
 View more versions of this product
Product Code. 16889783
  • £349.00 / 150µL
Explore more special offers
Description
Description
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2.
Specifications
Specifications
MARCH5
Polyclonal
Unconjugated
MARCH5
MARCH V, MARCH5, Mitochondrial ubiquitin ligase, MITOL, RING finger protein 153, RNF153
Rabbit
Antigen Affinity Chromatography
RUO
54708
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NX47
MARCH5
MARCH5 Fusion Protein Ag2806
150 μL