Learn More
Novus Biologicals™ MARCKS like protein Overexpression Lysate
Brand: Novus Biologicals™ NBL1-12896
Additional Details : Weight : 0.00970kg
Description
The MARCKS like protein Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
The lysate was created in HEK293T cells, using plasmid ID RC23181 and based on accession number NM_239. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens MARCKS-like 1 (MARCKSL1), mRNA. | |
Native |
For Research Use Only