missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | MARK1 |
---|---|
Concentration | 360 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16851145
|
Proteintech
21552-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16841145
|
Proteintech
21552-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The MARK1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human MARK1. This antibody recognizes human, mouse, rat antigen. The MARK1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
MARK1 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
360 μg/mL | |
Polyclonal | |
Rabbit | |
4139 | |
Primary |